pH Indicator solution, wide range pH 4-10

Supplier: Hach
Danger

2329332
97026-686EA 55.5 USD
97026-686 89231-042 104086-120
pH Indicator solution, wide range pH 4-10
pH Indicator solution

For visual determination of pH, using test kits


Method: 10222

UN: 1219
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR