Bromocresol green-methyl red solution pH-indicator
Supplier: Hach
Synonyms: Methyl Red - Bromocresol green solution
2329232 2329249
97026-600EA 51.07 USD
97026-600 89231-038 89231-040 104086-116 104086-118
Bromocresol green-methyl red solution pH-indicator
Bromocresol green-methyl red
pH indicator for alkalinity determination by titration
APHA formulation.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.