ISE Standards
Supplier: Hach
1279249 2351749 2354153 1474949 23249 2370853 2835153 30749 2406549
104085-274EA 87.61 USD
104085-274 104086-140 104086-144 104085-546 104086-114 104086-160 104086-702 104086-754 104088-210
ISE Standards
Standards Environmental Analysis Standards
ISE standards are used to calibrate ion selective electrode measurement systems by coresponding the electrode output with the concentration of the analyte. Pairs of standards with concentrations that differ by a factor of ten are used for analysis.