pH Buffer Solutions, Color Coded

Supplier: Hach

2283661 2283456 2283461 2283526 2283536 2283556 2283561 2283626 2283656 2283426 2283436 2947600
104086-080EA 387.39 USD
104086-080 104086-060 104086-062 104086-064 104086-066 104086-070 104086-072 104086-074 104086-078 104086-054 104086-056 104086-722
pH Buffer Solutions, Color Coded
Buffers pH Calibration Buffers

  • For calibration of pH meters
  • Color coded for easy identification
  • Accuracy: pH is ±0.02 at 25 °C


Certifications: Traceable to NIST.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR