Titrator, Digital
Supplier: Hach
2271900
104086-030EA 209.9 USD
104086-030
Titrator, Digital
Titrators
Portable and robust digital Titrator for fast and precise titrations. High resolution (125 µl/digit) and high accuracy (±1%). Applications for more than 40 parameters with interchangeable cartridges.
  • Digital Titrator methods use a high-precision dispensation device and cartridges of concentrated titrant to allow analyses in the lab, the plant or the field
  • Interchangeable cartridges allow you to change titrants in seconds; no cleaning or rinsing burettes
  • Versatile: More than 50 parameter/range combinations are available
  • Concentrated titrants allow 40 to 100 typical titrations per cartridge
  • End-Point easier to determine: The number used for calculation of sample concentration is recorded directly from the Digital Titrator's digital counter; for many methods, the displayed value is the concentration
Sulfite Test Kit, Model SU-DT
- Kit contains 100 tests with a range of 10 to 800 mg/L
- Digital Titrator and standardized titration cartridges provide laboratory accuracy for field titrations
- Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field
Hardness (Total & Calcium) Test Kit, Model HAC-DT
- Range: 10 to 4000 mg/L
- Contains approximately 100 of each (Total and Calcium) test
- Digital titrator offers higher levels of precision and accuracy than drop count procedures, with reproducibility comparable to a burette titration
- Rugged titration device is ideal for field use, with no glass burets required
Alkalinity Reagent Set (without titrator)
- For alkalinity determination by sulphuric acid titration with the digital titrator
- Range: 10 to 4000 mg/L CaCO₃
- Approximately 40 to100 tests
- Method: 8203 (Phenolphthalein and Total Alkalinity)