Soil Extractant Solution, Mehlich 2
Supplier: Hach
2266349
104086-016EA 62.43 USD
104086-016
Soil Extractant Solution, Mehlich 2
Environmental Test Kits
The Mehlich 2 extractant is used for phosphate and potassium determination in soils.