Nitrogen-Nitrate Reagent Set, LR, 5 ml Sample, Hach
Supplier: Hach
2245100 2429800
89231-094EA 122 USD
89231-094 104085-996 104086-228
Nitrogen-Nitrate Reagent Set, LR, 5 ml Sample, Hach
Environmental Test Kits
For determination of Nitrogen-Nitrate by the Cadmium Reduction method
Sample Size: 5 mL For visual test kits. Reagent set includes: NitriVer® 3 Powder Pillows and NitraVer® 6 Reagent Powder Pillows for 100 tests.