Colour Test Kit
Supplier: Hach
223400
104085-958EA 253.77 USD
104085-958
Colour Test Kit
Environmental Test Kits
Color disc kit reads apparent Color created by dissolved substances that can indicate industrial, agricultural or natural pollution. Dual range measurement in Color units.
Delivery information: Includes Color disc, comparator box and tubes, instructions and carrying case.