Chromium Test Kits, Comparator Method, Models CH-8 and CH-12
Supplier: Hach
222800
104085-946EA 463.85 USD
104085-946
Chromium Test Kits, Comparator Method, Models CH-8 and CH-12
Environmental Test Kits
These kits are used to determine chromium or chromium(VI) using simple, economical color disc methods.
Delivery information: CH-12 kit includes four reagents, clippers, two colour discs, comparator box, two viewing tubes, bottle, flask, dropper tube, stove and cover, test tube clamp, instructions and carrying case. CH-8 kit includes one reagent, two colour discs, comparator box, two viewing tubes, vial, instructions and carrying case.