Buffer Powder Pillows
Supplier: Hach
1407995
1409895
1410766
2227166
2227195
2226964
2226966
2226995
2227064
2227066
2227095
2227164
2299264
104085-326PK
35.81
USD
104085-326
104085-334
104085-338
104085-942
104085-944
104085-926
104085-928
104085-930
104085-934
104085-936
104085-938
104085-940
104088-198
Buffer Powder Pillows
Buffers
pH Calibration Buffers
The unit-dose powder pillows assure buffer freshness and eliminate the possibility of contamination. Each powder pillow prepares 50 ml of buffer solutions of respective pH.
- Fresh and reliable
- No danger of contamination
- Traceable to NIST standards
Delivery information: The buffer pillows are delivered in an airtight container and are supplied with a certificate of conformity.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...