Visual Comparator Test Kit, Ethylene Glycol
Supplier: Hach
2186400
104085-912EA
366.74
USD
104085-912
Visual Comparator Test Kit, Ethylene Glycol
Environmental Test Kits
Ethylene glycol (anti-freeze) contamination of automotive lubricants is an indication of a cracked engine block. Glycol presence also can indicate leaks in cooling systems. This test kit model EG-1 presents a purple color if ethylene glycol and other 1,2 glycols are present in oil or water.
Delivery information: Kit includes sample pipettes, vials and reagents for 25 tests, standard solution, instructions and carrying case.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)