Nitrites Test Kits, NI-15
Supplier: Hach
2182000
104085-892PK 274.72 USD
104085-892
Nitrites Test Kits, NI-15
Environmental Test Kits
The accurate visual test kit method to determine nitrogen as nitrite by diazotisation of sample followed by color comparison.
  • Simple and cost effective
  • Portable test kit
Delivery information: Kit includes color disc, comparator box and tubes, reagents (but not deionized water), instructions and carrying case.