Secchi Disc
Supplier: Hach
2177200
104085-888EA 204.58 USD
104085-888
Secchi Disc
Turbidity Meters
The disc is designed to measure water transparency in situ via the semi-quantitative method.
  • Disc slowly lowered in the water until the pattern is no longer visible
  • Depth of immersion is then evaluated
  • Measure related to the transparency
For use in oceans and lakes.
Ø = 200 mm
Certifications: ISO 7027.