1-(2-Pyridylazo)-2-naphthol (PAN) solution 0.3% indicator

Supplier: Hach
Danger

Synonyms: PAN, 1-(2-Pyridylazo)-2-naphthol

2150232
97026-648EA 107.26 USD
97026-648 104085-874
1-(2-Pyridylazo)-2-naphthol (PAN) solution 0.3% indicator
1-(2-Pyridylazo)-2-naphthol

For low-level determination of nickel and cobalt by the PAN method


Hach method 8078.

Formula: C₁₅H₁₁N₃O
MW: 249.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004071
CAS Number: 85-85-8
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR