Phthalate-Phosphate Reagent powder Pillows
Supplier: Hach
2150166
104085-872PK 81.31 USD
104085-872
Phthalate-Phosphate Reagent powder Pillows
Environmental Test Kits
For low-level determination of nickel and cobalt by the PAN method.
Packaging: Pack of 50 powder pillows.