Benzotriazole

Supplier: Hach
Danger

Synonyms: 1,2,3-Benzotriazole, 1H-1,2,3-Benzotriazole, 1H-Benzotriazole, 1H-Benzo[d][1,2,3]triazole

2141342
104085-866EA 140.49 USD
104085-866
Benzotriazole
Benzotriazole
Formula: C₆H₅N₃
MW: 119.13 g/mol
Boiling Pt: 350
Melting Pt: 99
Flash Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00005699
CAS Number: 95-14-7
EINECS: 202-394-1
UN: 2811
ADR: 6.1 III
Merck Index: 13,01109
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ba54cc42ef5ad9dbae6d7f118a3af1ae642287&vid=ce57b3f0d2583e32220d1e191d0202ad452e6475&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR