Lauryl Tryptose Broth MPN Tubes, Concentrated
Supplier: Hach
2101415
104085-764PK 85 USD
104085-764
Lauryl Tryptose Broth MPN Tubes, Concentrated
Media for Microbiology Prepared Media for Microbiology
Lauryl tryptose broth MPN tubes, concentrated. Selectivity: total coliform bacteria (presumptive).
  • Save time and labor costs with ready to use MPN tubes, with minimal training requirements