Iron(III) chloride solution in sulfuric acid

Supplier: Hach
Danger

204253
89230-928PK 28.13 USD
89230-928 10820-126 104085-710
Iron(III) chloride solution in sulfuric acid
Iron (III) chloride

Ferric Chloride-Sulfuric Acid Solution for determination of volatile acids by the esterification method.

Formula: FeCl₃
MDL Number: MFCD00011005
CAS Number: 7705-08-0
UN: 3264
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR