Sodium hydroxide 4.5 N volumetric solution

Supplier: Hach
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

204053
89230-926EA 69.8 USD
89230-926 104085-708
Sodium hydroxide 4.5 N volumetric solution
Sodium hydroxide

For volatile acids determination by the esterification method.


Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR