Sulfuric acid 0.100 N volumetric solution

Supplier: Hach
Danger

20253
104085-696EA 59.56 USD
104085-696
Sulfuric acid 0.100 N volumetric solution
Sulfuric acid

Standardized acid titrating solution. Used for various titration procedures and pH adjustments.

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
MDL Number: MFCD00064589
CAS Number: 7664-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR