Water Quality Test Discs, HA-77
Supplier: Hach
202300
104085-694KT 324.36 USD
104085-694
Water Quality Test Discs, HA-77
Environmental Test Kits
Hardness and iron determination in a water quality test kit which offers accuracy and portable utility in one package. Utilising drop count titration (hardness) and color disc colorimetry.
  • Easy to use
  • Portable kit
Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Delivery information: Includes four reagents, bottles, flask, color discs, comparator box, viewing tubes, instructions and carrying case sufficient for 100 tests.