Test Kit, Free Cyanide
Supplier: Hach
201002
104085-692PK 563.09 USD
104085-692
Test Kit, Free Cyanide
Environmental Test Kits
The test kit is suitable for measuring the effectiveness of cyanide removal from waste effluents of metal-cleaning and electroplating baths and various other chemical processes through alkaline or neutral chlorination procedures. Not suitable for complexed cyanides.
  • Simple and fast color comparison method
  • Portable test kit for on site measurement
Delivery information: Test kit comprises reagents as powder pillows, comparator, comparator disc, tubes and simple equipment for sample treatment and measurement, all in a sturdy carrying case.