TenSette® Single Channel Pipettes, Variable Volume
Supplier: Hach
1970001 1970010
104085-676EA 936.68 USD
104085-676 104085-678
TenSette® Single Channel Pipettes, Variable Volume
Pipettes Micropipettes
With these pipettes users are able to perform routine laboratory pipetting quickly, economically, and accurately. The pipettes dispense from 0.1 to 1 ml or from 1 to 10 ml in 0.1 or 1.0 ml increments.
  • Save time, reduces glassware cleanup and assures accuracy
  • Specifically designed for use with Hach Voluette and PourRite standards in Standard Additions accuracy checks
Ordering information: Set includes pipette, manual and 50 disposable tips.