Hexamethylenetetramine, powder pillow buffer substance

Supplier: Hach

187866
97026-346PK 78.59 USD
97026-346 104085-644
Hexamethylenetetramine, powder pillow buffer substance
Methenamine

For aluminum determination by the Eriochrome Cyanine R method (Hach method 8326)


Sample Size: 25 mL.


Ordering information: Pack of 50 powder pillows.

Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.

Formula: C₆H₁₂N₄
Storage Temperature: Ambient
MDL Number: MFCD00006895
CAS Number: 100-97-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3ecbb5df5394322c2150525ce6e9f48b5aadde7&vid=04b623f7f333c7cf159016619cf91fc792af9a3f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR