Test Kits, Cyanuric Acid
Supplier: Hach
185102
104085-642EA 274.15 USD
104085-642
Test Kits, Cyanuric Acid
Environmental Test Kits
A stabilizing agent for chlorine, cyanuric acid is used in swimming pool treatment. Cyanuric acid test kit measures the turbidity formed in proportion to cyanuric acid concentration, after the reagent is added.
Delivery information: Includes one reagent, clippers, graduated cylinder, sample cell with cap, dipstick instructions and carrying case.