Multiparameter Test Kits, HA-62
Supplier: Hach
183700 183701 183702
104085-636EA 464.15 USD
104085-636 104085-638 104085-640
Multiparameter Test Kits, HA-62
Environmental Test Kits
A multi-parameter test kit for hardness, iron and pH, which provides an easy-to-use, cost effective way to determine the quality of water.
  • Popular tests in one rugged kit
  • Uses color disc and drop count titration methods