Sulphide Reagents 1 and 2
Supplier: Hach
181632 181732 181749
104085-622EA 63.79 USD
104085-622 104085-626 104085-628
Sulphide Reagents 1 and 2
Environmental Test Kits
Used for sulphide determination by the methylene blue method.
  • Range upto 800 µg/L
  • Used with visual test kits