Ferroin (1,10-phenanthrolineferrous sulfate complex) solution indicator, for COD determination

Supplier: Hach

Synonyms: Tris(1,10-phenanthroline)iron (II) sulphate, Tris(1,10-phenanthroline-N1,N10)iron sulphate, Tris(1,10-phenanthroline)iron (II) sulfate, Tris(1,10-phenanthroline-N1,N10)iron sulfate

181233
104085-616EA 44.42 USD
104085-616
Ferroin (1,10-phenanthrolineferrous sulfate complex) solution indicator, for COD determination
Ferroin (1,10-Phenanthroline-ferrous sulphate-complex)

Oxidation-reduction indicator solution used in determination of COD by ferrous titration.

Formula: C₃₆H₂₄FeN₆SO₄
MW: 692.54 g/mol
MDL Number: MFCD00036428
CAS Number: 14634-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR