Potassium iodide

Supplier: Hach

Synonyms: Dipotassium diiodide, Kali iodide

16726H
104085-596EA 141.59 USD
104085-596
Potassium iodide
Potassium iodide
Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330
Melting Pt: 686
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR