Phenolphthalein 5 g/L diethylene glycol monoethyl ether APHA indicator for titration

Supplier: Hach
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

16232
97026-170EA 38 USD
97026-170 10000-782 104085-590
Phenolphthalein 5 g/L diethylene glycol monoethyl ether APHA indicator for titration
Phenolphthalein

pH indicator for use in titrations


Color change: colorless to pink at ~pH 8.3.

Formula: C₂₀H₁₄O₄
MW: 318.33 g/mol
MDL Number: MFCD00005913
CAS Number: 77-09-8
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR