tri-Sodium citrate dihydrate

Supplier: Hach

1490849
104085-578EA 55.58 USD
104085-578
tri-Sodium citrate dihydrate
tri-Sodium citrate dihydrate
Formula: Na₃C₆H₅O₇·2H₂O
Melting Pt: 150 °C
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR