Orthoboric acid, powder pillow for titrimetry

Supplier: Hach

1481799
97026-622PK 143.87 USD
97026-622 104085-564
Orthoboric acid, powder pillow for titrimetry
ortho-Boric acid

For Ammonia determination by distillation.


Ordering information: Pack of 100 Powder Pillows.

Formula: B(OH)₃
MDL Number: MFCD00011337
CAS Number: 10043-35-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR