Magnesium chloride 510 g/L in aqueous solution

Supplier: Hach

1476253
104085-550EA 175.02 USD
104085-550
Magnesium chloride 510 g/L in aqueous solution
Magnesium chloride
Formula: MgCl₂
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR