Visual Comparator Test Kits, Nitrate, NI-11
Supplier: Hach
146803
104085-526EA 274.72 USD
104085-526
Visual Comparator Test Kits, Nitrate, NI-11
Environmental Test Kits
Simple, low cost, portable, color disc method of testing nitrate.
Delivery information: Kit includes color disc, comparator box and tubes, reagents, instructions and carrying case.