Visual Comparator Test Kits, Iron
Supplier: Hach
146401 2299300 2667200 146500 255600 146400
104085-520EA 190.69 USD
104085-520 104086-090 104086-562 104085-522 104086-402 104088-114
Visual Comparator Test Kits, Iron
Environmental Test Kits
Qualitative results using traditional color comparator disc methods.
  • Portable - ideal for field testing
  • Simple fast method
  • Kits contain all required reagents and apparatus in a rugged carrying case for 100 tests
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.