LDPE Stoppers, Hollow
Supplier: Hach
1448000
104085-474PK 40.35 USD
104085-474
LDPE Stoppers, Hollow
Stoppers PE/LDPE/HDPE Stoppers
Ideally can be used as a small beaker, sample cup for pH meters, flame photometers and automatic analyzers.
  • LDPE
  • Interchangeable with rubber stoppers
  • Safer and economical
  • Fits snugly