Sodium hydroxide 0.3636 N, cartridge for digital titrator

Supplier: Hach
Danger

1437801
97026-240EA 61.65 USD
97026-240 104085-434
Sodium hydroxide 0.3636 N, cartridge for digital titrator
Sodium hydroxide

Used for carbon dioxide determination by Digital Titration.


  • Accurate results - when and where you need them
  • Fast and Flexible
  • Versatile
  • Economical


Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.

Formula: NaOH
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Method 8205
Method Name Phenolphthalein
Number of tests 40 - 100
Parameter Carbon Dioxide
Platform Digital Titrator
Range 10 - 100 mg/L CO2
Titrant 0.3636 N NaOH


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR