Acid Reagent Powder Pillows for Chromium, Hach
Supplier: Hach
220399 2603549 2603449 1421199 2182369 1432399 1436369 107266 1486266 1261699 1416066 1403268 183699 2615199 94799 1429449 1432198 1432298 2081599 2118869 2106669 2167969 87299 2395266 2395466 96299 85299 2481569 2380268 1486166 1486398 2244100 2254069 94299 94399 2107669
10000-812PK 108.32 USD
10000-812 MSPP-220399 MSPP-2603549 MSPP-2603449 104085-386 104085-896 104085-422 104085-426 104085-222 104085-572 104085-262 104085-356 104085-288 104085-634 104086-486 104087-082 104085-414 104085-418 104085-420 104085-744 104085-834 104085-796 104085-880 104086-988 104086-184 104086-186 104087-132 104086-972 104086-332 104086-178 104088-118 104088-120 104088-188 104088-190 104088-370 104088-372 104088-160
Acid Reagent Powder Pillows for Chromium, Hach
Environmental Test Kits
For total chromium determination by the alkaline hypobromite oxidation method.