Potassium Iodide-Iodate Standard Solution, 0.0125 N
Supplier: Hach
1400153
104085-286EA 115.7 USD
104085-286
Potassium Iodide-Iodate Standard Solution, 0.0125 N
Standards Environmental Analysis Standards
Used for determination of sulphite by burette iodometric titration.