Silicone oil

Supplier: Hach

Synonyms: Polyphenyl-methylsiloxane

126936
104085-266EA 36.55 USD
104085-266
Silicone oil
Silicone oil

Multiple uses, including preparation of turbidity sample cells.

Storage Temperature: Ambient
MDL Number: MFCD00132673
CAS Number: 68083-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR