Silver Nitrate Standard Solution
Supplier: Hach
1255149
104085-260EA 120.01 USD
104085-260
Silver Nitrate Standard Solution
Standards Environmental Analysis Standards
Standard solution for chloride determination by argentometric burette titration.