Powder Pillows, Chloride 2 Indicator
Supplier: Hach
104399 105766
104085-206PK 61.85 USD
104085-206 104085-216
Powder Pillows, Chloride 2 Indicator
Environmental Test Kits
Powder reagent in sealed package used as indicator for the argentometric titration of chloride.
Caution: Note: Product contains potsassium chromate. Please read MSDS prior to usage.