Amidosulfonic acid, powder pillow

Supplier: Hach
Warning

Synonyms: Aminosulphonic acid, Amidosulphuric acid, Sulfamic acid, Sulfamidic acid, Amidosulphuric Acid, Sulphamic acid, Amidosulphonic acid, Sulphamidic acid, Aminosulfonic acid, Amidosulfonic acid, Amidosulfuric Acid, Amidosulfuric acid

105599
97026-176PK 47.72 USD
97026-176 104085-214
Amidosulfonic acid, powder pillow
Amidosulphonic acid

Acid reagent used in chlorine, chromate, and sulfite determinations.


Pack of 100 Powder Pillows.

Formula: H₃NO₃S
MW: 97.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011603
CAS Number: 5329-14-6
UN: 2967
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afa0500786be69d531cdf06f8e67548e6f67b23f&vid=73aa20196fbabf965e4375f2cf3675e2f513ee59&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR