Sodium metaarsenite solution 5.0 g/L

Supplier: Hach

104732
97026-662EA 40.8 USD
97026-662 104085-212
Sodium metaarsenite solution 5.0 g/L
Sodium metaarsenite

For sample dechlorination prior to analysis.

Formula: NaAsO₂
MDL Number: MFCD00003472
CAS Number: 7784-46-5
UN: 2027
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR