Sodium hydroxide 1.000 volumetric solution

Supplier: Hach
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

104532
10765-202EA 47.63 USD
10765-202 104085-208
Sodium hydroxide 1.000 volumetric solution
Sodium hydroxide

Multi-purpose sodium hydroxide solution for pH adjustment.

Formula: NaOH
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR