Anti-NNT Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-5097R-CY7
10408-150EA 823.02 USD
10408-150
Anti-NNT Rabbit Polyclonal Antibody (Cy7®)
Antibodies
The transhydrogenation between NADH and NADP is coupled to respiration and ATP hydrolysis and functions as a proton pump across the membrane. May play a role in reactive oxygen species (ROS) detoxification in the adrenal gland.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NNT
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR