Anti-SLCO2A1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-4710R-CY3
10405-654EA 823.27 USD
10405-654
Anti-SLCO2A1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
May mediate the release of newly synthesized prostaglandins from cells, the transepithelial transport of prostaglandins, and the clearance of prostaglandins from the circulation. Transports PGD2, as well as PGE1, PGE2 and PGF2A.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLCO2A1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR