Anti-HIV1 p55+p17 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-4982R-CY3
10405-288EA 823.01 USD
10405-288
Anti-HIV1 p55+p17 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
HIV1 performs highly complex orchestrated tasks during the assembly, budding, maturation and infection stages of the viral replication cycle. During viral assembly, the proteins form membrane associations and self-associations that ultimately result in budding of an immature virion from the infected cell. Gag precursors also function during viral assembly to selectively bind and package two plus strands of genomic RNA. HIV1 Gag p17, the HIV1 matrix protein is produced by the digestion of its precursor Gag p55 by HIV1 protease.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HIV1 p55+p17
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR