Locking Clips, with Breakaway Lifting Tabs
Supplier: Alkali Scientific
MT2086
104029-088EA 38.45 USD
104029-088
Locking Clips, with Breakaway Lifting Tabs
Clamps General Purpose Clamps
Secure tubes with ease using Stop-Pop™ Locking clips, designed for 1.5 to 2 ml tubes.
  • Prevents 1.5 to 2 ml minicentrifuge tubes from popping open during heating/boiling
Secure sample tubes with ease using the Stop-Pop™ Locking clips. These locking clips feature breakaway lifting tabs for convenient tube retrieval. The innovative design ensures a tight seal to prevent sample leakage or contamination during storage or transport. Each pack contains 100 locking clips, providing an efficient solution for tube closure in laboratory workflows.