Radiant™ 1-Step RTPCR Kit
Supplier: Alkali Scientific
QRT201
104029-064EA 101.52 USD
104029-064
Radiant™ 1-Step RTPCR Kit
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Conduct efficient reverse transcription PCR with Alkali Scientific™ Radiant™ 1-Step RTPCR Kit, offering 50 reactions for convenient gene expression analysis.
  • Obtain high sensitivity and yield in a wide amplification range: Detect as little as 1 fg of total RNA
  • Achieve high specificity and reduce primer-dimers
  • Count on excellent performance with the kit′s proprietary hot start buffer
  • Produce long fragments: Generate transcripts up to 6.5 kb
  • Transcribe a variety of templates, even the most difficult (e.g., GC-rich RNA): Overcome high secondary structure with thermostable transcriptor reverse transcriptase in combination with an optimized buffer
  • Save time and improve results with a convenient kit: Minimize pipetting steps and prevent contamination by using a pre mixed enzyme blend and optimized buffer that includes PCR-grade dNTPs and hot start components
  • Safeguard your RNA from degradation by RNases with the supplied thermostable protector RNase inhibitor that is functional during one-step RT-PCR when other RNase inhibitors fail
  • Increase confidence in your results: Use the supplied control reactions to monitor performance
Empower your molecular biology research with the Radiant™ 1-Step RT-PCR Kit. Specifically designed for sensitive and efficient reverse transcription polymerase chain reaction (RT-PCR) applications, this kit offers a streamlined workflow and reliable performance. With enough reagents for 50 reactions, it provides a cost-effective solution for high-throughput experiments. The kit combines reverse transcription and PCR amplification into a single step, minimizing handling steps and reducing the risk of contamination. Achieve accurate and reproducible results with the Radiant™ 1-Step RT-PCR Kit, accelerating your research progress and enabling discoveries in gene expression analysis, viral detection and RNA quantification.